Learn More
Cadmium chloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 296330050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
960.0°C | |
568.0°C | |
10108-64-2 | |
Powder | |
99.99% | |
CdCl2 | |
04,485 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
dichlorocadmium | |
24947 | |
183.31 | |
Trace Metal Basis |
Cadmium chloride | |
99.99% | |
White | |
5 g | |
CdCl2 | |
MFCD00010916 | |
15, 1622 | |
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether | |
[Cl-].[Cl-].[Cd++] | |
183.31 | |
CHEBI:35456 | |
99.99% | |
Glass bottle |
Chemical Identifiers
10108-64-2 | |
183.31 | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
24947 | |
dichlorocadmium |
CdCl2 | |
MFCD00010916 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
CHEBI:35456 | |
[Cl-].[Cl-].[Cd++] |
Safety and Handling
GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S
GHS Signal Word: Danger
EINECSNumber : 233-296-7
RTECSNumber : EV0175000
TSCA : TSCA
RUO – Research Use Only