missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium chloride, 98%, pure, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC192695000

 View more versions of this product

Catalog No. AC192695000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cadmium chloride
97.5% min. (Argentometry)
CdCl2
15,1622
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
Cl[Cd]Cl
183.31
CHEBI:35456
98%
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
960.0°C
500g
10108-64-2
CdCl2
04,485
caddy, cadmium chloride, cadmium dichloride, cadmium ii chloride, caswell no. 135, ccris 114, hsdb 278, kadmiumchlorid, vi-cad
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.31
Pure
0.05% max
568.0°C

Chemical Identifiers

10108-64-2
183.31
caddy, cadmium chloride, cadmium dichloride, cadmium ii chloride, caswell no. 135, ccris 114, hsdb 278, kadmiumchlorid, vi-cad
CHEBI:35456
Cl[Cd]Cl
CdCl2
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
Safety and Handling

Safety and Handling

GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S

GHS Signal Word: Danger

EINECSNumber : 233-296-7

SDS Documents