missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium carbonate, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1186414

 View more versions of this product

Catalog No. AA1186414



Specifications

Specifications

Cadmium carbonate
99.99%
MFCD00010918
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
C(=O)([O-])[O-].[Cd+2]
172.422
172.42
4.26g/mL
White
513-78-0
CCdO3
White powder/lumps
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+);carbonate
10564
Powder/Lump
Odorless
25g

Chemical Identifiers

513-78-0
172.422
GKDXQAKPHKQZSC-UHFFFAOYSA-L
10564
C(=O)([O-])[O-].[Cd+2]
CCdO3
MFCD00010918
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
cadmium(2+);carbonate
Safety and Handling

Safety and Handling

H400-H410-H302-H312-H332

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 208-168-9

Documents