missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium carbonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A158120B

 View more versions of this product

Catalog No. AAA158120B



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cadmium carbonate
513-78-0
CCdO3
UN2570
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
[Cd++].[O-]C([O-])=O
172.42
172.42
Odorless
500°C (decomposition)
1000 g
MFCD00010918
14,1616
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+);carbonate
10564
97%
4.26

Chemical Identifiers

513-78-0
172.42
GKDXQAKPHKQZSC-UHFFFAOYSA-L
10564
CCdO3
MFCD00010918
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
[Cd++].[O-]C([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H302+H312-H330-H340-H350-H361-H372

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 208-168-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only