Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium bromide, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Cadmium bromide
CAS 7789-42-6
Name Note Ultra dry
Formula Weight 272.22
MDL Number MFCD00015996
UN Number UN2570
Merck Index 14,1615
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4710506

  SDS
alfa aesar™
4710506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4710514

  SDS
alfa aesar™
4710514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 272.22
Packaging Ampule
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to pale yellow
Chemical Name or Material Cadmium bromide
Density 5.192g/mL
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
MDL Number MFCD00015996
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
TSCA TSCA
UN Number UN2570
Quantity 5g
Odor Odorless
Assay 99.999%
CAS 7789-42-6
Boiling Point 863°C
Melting Point 567°C
Sensitivity Hygroscopic
RTECS Number EU9935000
Name Note Ultra dry
Merck Index 14,1615
Physical Form Powder
Assay Percent Notes (metals basis)
Storage Note 1
  • Ampouled under argon

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

  • Ampouled under argon