missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium bromide hydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4449836

 View more versions of this product

Catalog No. AA4449836



Specifications

Specifications

Cadmium bromide hydrate
(metals basis)
MFCD00150204
cadmium 2+ dibromide
Crystalline
Odorless
White to Yellow
681435-25-6
CdBr2·xH2O
UN2570
272.22 (Anhydrous)
99.999%
Hygroscopic
500g

Chemical Identifiers

681435-25-6
CdBr2·xH2O
MFCD00150204
cadmium 2+ dibromide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

EINECSNumber : 232-165-1

RTECSNumber : EU9935000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents