missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium bromide hydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4449822

 View more versions of this product

Catalog No. AA4449822



Specifications

Specifications

Cadmium bromide hydrate
(metals basis)
MFCD00150204
cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
Crystalline
Odorless
White to Yellow
7789-42-6
CdBr2·xH2O
UN2570
272.22 (Anhydrous)
99.999%
Hygroscopic
100g

Chemical Identifiers

7789-42-6
CdBr2·xH2O
MFCD00150204
cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EU9935000

TSCA : TSCA

SDS Documents