Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium bromate hydrate, 98%

Chemical Identifiers

Chemical Name or Material Cadmium bromate hydrate
CAS 14518-94-6
Molecular Formula Br2CdO6·xH2O
Linear Formula Cd(BrO3)2·xH2O
Formula Weight 368.21 (Anhydrous)
MDL Number MFCD00015991
UN Number UN1450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1796414

  SDS
alfa aesar™
A1796414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 368.21 (Anhydrous)
Chemical Name or Material Cadmium bromate hydrate
Assay 98%
Molecular Formula Br2CdO6·xH2O
Linear Formula Cd(BrO3)2·xH2O
MDL Number MFCD00015991
CAS 14518-94-6
UN Number UN1450
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H272-H302-H312-H332
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P221-P210-P220-P261-P304+P340-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BROMATES, INORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P221-P210-P220-P261-P304+P340-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H302-H312-H332
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.