Please login to your online account to display your discounted pricing

Cadmium Acetate, Dihydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$412.09 - $412.09

Chemical Identifiers

Chemical Name or Material Cadmium Acetate
CAS 5743-04-4
Name Note Dihydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula (CH3COO)2Cd·2H2O
Formula Weight 266.52
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002495

 
avantor performance
119004
Poly Bottle 125g Each for $412.09
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 266.52
Chemical Name or Material Cadmium Acetate
CAS 5743-04-4
Name Note Dihydrate
Quantity 125g
Specific Gravity 1L = 2.01kg
Packaging Poly Bottle
Molecular Formula (CH3COO)2Cd·2H2O
Purity Grade BAKER ANALYZED™ Reagent