missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium acetate dihydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 317131000

 View more versions of this product

Catalog No. AC317131000


 

Specifications Description & Specifications

Specifications

≥97.5%
266.52
White
50ppm max.
500ppm max.
30ppm max.
C4H6CdO4·2H2O
MFCD00150020
10ppm max.
Plastic bottle
5743-04-4
Crystals or Crystalline Powder
100g
For analysis
50ppm max.
(CH3CO2)Cd·2H2O
Cadmium acetate dihydrate
10ppm max.
10ppm max.
0.005% max.
0.1% max.
98%

Chemical Identifiers

5743-04-4
C4H6CdO4·2H2O
266.52
Cadmium acetate dihydrate
For analysis
98%
(CH3CO2)Cd·2H2O
MFCD00150020
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Very toxic to aquatic life with long lasting effects. May cause cancer. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Obtain special instructions before use. Use personal protective equipment as required.

Danger