Please login to your online account to display your discounted pricing

Cadmium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Cd), LabChem™


Quantity 500mL
Packaging Poly Bottle

Manufacturer: lab chem  LC126001

 View more versions of this product

Catalog No. LC126001

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 112.4
Purity Grade Certified Grade
Linear Formula Cd
Traceability to NIST Traceable to NIST
CAS Max % 95.3
Concentration 1000ppm ±10ppm (1mL = 1mg Cd)
Chemical Name or Material Cadmium AA Standard
CAS Max % 4.6
Name Note 1000ppm (1mL = 1mg Cd)
UN Number UN3264
Recommended Storage Room Temperature
Physical Form Liquid
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Collect spillage.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Health Hazard 2 GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Very toxic to aquatic life.
Solubility Soluble in water
Density 1g/mL
CAS 7732-18-5
Identification Passes Test
CAS 7440-43-9
CAS 7697-37-2
CAS Max % 0.1
Quantity 500mL
Color Clear, colorless
Health Hazard 1 Danger
Packaging Poly Bottle

Specifications

Chemical Name or Material Cadmium AA Standard
CAS 7732-18-5
CAS Max % 95.3
CAS 7697-37-2
CAS Max % 4.6
CAS 7440-43-9
CAS Max % 0.1
Name Note 1000ppm (1mL = 1mg Cd)
Purity Grade Certified Grade
Linear Formula Cd
Formula Weight 112.4
UN Number UN3264
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Collect spillage.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Very toxic to aquatic life.

Room Temperature