Please login to your online account to display your discounted pricing

Cadmium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Cd), LabChem™

$50.63 - $94.51

Chemical Identifiers

Chemical Name or Material Cadmium AA Standard
CAS 7732-18-5
CAS Max % 95.3
CAS 7697-37-2
CAS Max % 4.6
CAS 7440-43-9
CAS Max % 0.1
Name Note 1000ppm (1mL = 1mg Cd)
Purity Grade Certified Grade
Linear Formula Cd
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC126001

 
lab chem
LC126001
Poly Bottle 500mL Each for $94.51

LC126007

 
lab chem
LC126007
Poly Bottle 125mL Each for $50.63
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 112.4
Chemical Name or Material Cadmium AA Standard
Color Clear, colorless
Quantity 500mL
CAS Max % 0.1
Name Note 1000ppm (1mL = 1mg Cd)
Physical Form Liquid
Concentration 1000ppm ±10ppm (1mL = 1mg Cd)
Linear Formula Cd
CAS 7732-18-5
UN Number UN3264
Health Hazard 1 Danger
Identification Passes Test
CAS Max % 95.3
Health Hazard 2 GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Very toxic to aquatic life.
CAS 7697-37-2
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Collect spillage.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Specific Gravity 1g/mL
CAS Max % 4.6
Recommended Storage Room Temperature
Density 1g/mL
CAS 7440-43-9
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Collect spillage.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Very toxic to aquatic life.

Room Temperature