Please login to your online account to display your discounted pricing

Cadmium AA Standard, 1mL = 1mg Cd (1,000ppm Cd) in 3% HCl, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Cadmium AA Standard
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 7647-01-0
CAS Min % 3.88
CAS Max % 4.04
CAS 7440-43-9
CAS Min % 0.09
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ACD1KH-100

 
ricca chemical company
RIACD1KH100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACD1KH-500

 
ricca chemical company
RIACD1KH500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
CAS 7732-18-5
CAS Min % 94.0
Concentration or Composition Notes 1,000ppm Cd
Equivalence 1 mL = 1 mg Cd
CAS Min % 3.88
Packaging Poly natural
CAS Min % 0.09
CAS 7647-01-0
CAS Max % 4.04
CAS Max % 98.0
CAS Max % 0.1
CAS 7440-43-9
Chemical Name or Material Cadmium AA Standard