Please login to your online account to display your discounted pricing

Cadmium, >99% (metals basis), Alfa Aesar™

$1,224.15 - $1,224.15

Chemical Identifiers

Chemical Name or Material Cadmium
CAS 7440-43-9
Physical Form Coarse powder, 0.3-1.5mm for reductors
Assay Percent Range 99+% (Metals basis)
MDL Number MFCD00010914
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3996236

 
alfa aesar
39962-36
500g Each for $1,224.15
Description & Specifications

Specifications

Appearance Colorless to pale green coarse powder
Vapor Pressure 0.00013 hPa
Color Colorless to pale green
Quantity 500g
Chemical Name or Material Cadmium
CAS 7440-43-9
Physical Form Coarse powder, 0.3-1.5mm for reductors
Assay Percent Range 99+% (Metals basis)
MDL Number MFCD00010914
Health Hazard 1 H330-H350-H372-H341-H361-H400-H410; Fatal if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOT Information DOT Class: 6.1, Packaging Group: II
EINECS Number 231-152-8
Safety and Handling

H330-H350-H372-H341-H361-H400-H410; Fatal if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.