Please login to your online account to display your discounted pricing

Alfa Aesar™ Cadmium 2,4-pentanedionate, 98%

Chemical Identifiers

Chemical Name or Material Cadmium 2,4-pentanedionate
CAS 14689-45-3
Formula Weight 310.64
MDL Number MFCD00040412
UN Number UN2570
Synonym Cadmium acetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4196806

  SDS
alfa aesar™
4196806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4196814

  SDS
alfa aesar™
4196814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 310.64
Physical Form Powder
RTECS Number EV0260000
MDL Number MFCD00040412
UN Number UN2570
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Cadmium acetylacetonate
CAS 14689-45-3
Assay 98%
Chemical Name or Material Cadmium 2,4-pentanedionate
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.