missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cadherin, Pan, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cadherin, Pan, Rabbit Polyclonal Antibody.

$277.90 - $1,402.06

Specifications

Antigen Cadherin, Pan
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Host Species Rabbit
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB9036P Thermo Scientific™ Lab Vision™
RB9036P
Purified Ab with BSA and Azide 1mL Each for $1,402.06
RB9036P0 Thermo Scientific™ Lab Vision™
RB9036P0
Purified Ab with BSA and Azide 100μL Each for $277.90
RB9036P1 Thermo Scientific™ Lab Vision™
RB9036P1
Purified Ab with BSA and Azide 500μL Each for $839.47
RB9036R7 Thermo Scientific™ Lab Vision™
RB9036R7
Ready-to-use for IHC (UVLP) 7mL Each for $673.04
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cadherin, Pan, Rabbit Polyclonal Antibody.

Cadherins are calcium-dependent cell adhesion molecules, which play important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. In adhesion junctions, cadherins are bound to Beta and gamma catenins, which in turn bind to alpha catenin, an actin binding protein. Cadherins play important role in tumor invasion and metastasis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Cow and Xenopus.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of chicken N-cadherin

Molecular Weight: 120kDa

Positive Control: MCF-Cells, Tonsil and squamous epithelium

Cellular Localization: Membrane

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cadherin, Pan
Polyclonal
Rabbit
Junction Protein
A synthetic peptide from the C-terminus of chicken N-cadherin
Western Blot
Unlabeled
RUO
Primary
IgG