Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cadherin E, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cadherin E, Mouse Monoclonal Antibody.

$260.12 - $1,243.16

Specifications

Antigen Cadherin E/E-Cadherin
Clone SPM471
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS9470P

 
thermo scientific™ lab vision™
MS9470P
1mL Each for $1,243.16

MS9470P0

 
thermo scientific
MS9470P0
100µL Each for $260.12

MS9470P1

 
thermo scientific
MS9470P1
500µL Each for $850.29

MS9470R7

 
thermo scientific™ lab vision™
MS9470R7
7mL Each for $636.16

MS9470RQ

 
thermo scientific™ lab vision™
MS9470RQ
12mL Each for $692.08
Description & Specifications

Specifications

Antigen Cadherin E/E-Cadherin
Clone SPM471
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Host Species Mouse
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Recombinant protein encoding human Cadherin-E 600-707aa

Obtain accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cadherin E, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

Host Species: Mouse

Clone: SPM471

Isotype: IgG2b/κ

Species Reactivity: Human. Others not tested.

Epitope: 600-707aa

Immunogen: Recombinant protein encoding human Cadherin-E 600-707aa

Molecular Weight: 120kDa

Positive Control: Colon CA

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)