Please login to your online account to display your discounted pricing
Cadence Science™ One-way Stopcocks
Precision-machined, individually lapped, and carefully mated to maintain pressure characteristics
$117.29 - $403.53
Specifications
Product Type 1-way Stopcock
View More Specs
Products
Catalog Number Mfr. No. Connectors For Use With (Equipment) Material Price Quantity    
01-290-40
cadence science™
POP6021IND
1 female luer-lock and 1 male luer-lock Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $188.82
14-825-307
cadence science™
POP6020IND
Female luer to female luer Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $117.29
14-825-309
cadence science™
POP6021HPIND
1 threaded female luer-lock and 1 male luer-lock Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $403.53
14-825-310
cadence science™
POP6029IND
Male luer-lock to 6mm hose end Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $235.50
14-825-313
cadence science™
POP6027IND
8mm dia. hose end, 4mm dia. lumen Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $171.29
14-825-312
cadence science™
POP6026IND
6mm dia. hose end, 3mm dia. lumen Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $171.29
14-825-311
cadence science™
POP6030IND
1 male luer-lock and one 8mm hose end Other stopcocks, manifolds, adapters, tubing, needles and glass syringes Nickel Plated Brass Each for $235.50
Description & Specifications
Specifications
Product Type 1-way Stopcock
These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 PSI water pressure
  • High Pressure version is pressure resistant up to 20 bars (290 psi)