Cadence Science™ Nonflexible Stainless Steel Animal Feeding Needles
For feeding and dosing without injury to sensitive tissues.
Manufacturer:  Cadence Science™ POP7909
 View more versions of this product
Catalog No. 14-825-281
Specifications
Specifications
4mm
14G
7.62cm
Non-sterile
Animal Feeding Needle
No
3 in.
304 Stainless Steel
Animal feeding and dosing without injury to sensitive tissues, ideal for pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals
Documents