Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Manufacturer:  Cadence Science™ POP7975
 View more versions of this product
Catalog No. 14-825-17G
Description
Description
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
  • Available with or without a reinforcing bar
  • Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)
Specifications
Specifications
22 G
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar
Glass
Non-sterile
Reinforcing bar
2.87 in.
7.3cm
Micro Emulsifying Needle with Reinforcing Bar
Documents