Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Manufacturer:  Cadence Science™ POP7969
 View more versions of this product
Catalog No. 14-825-17A
Description
Description
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
  • Available with or without a reinforcing bar
  • Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)
Specifications
Specifications
16 G
Reinforcing bar
7.3 cm
Non-sterile
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar
2.87 in.
Glass
Micro Emulsifying Needle with Reinforcing Bar
Documents