Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
Manufacturer:  Cadence Science™ POP9923
 View more versions of this product
Catalog No. 14-825-251
Description
Description
  • Nontoxic, nonpyrogenic
  • PTFE tubing is flexible and softer than other types of tubing
  • Low friction protects delicate organs
  • Translucent for easy confirmation of reagent delivery
  • Plastic luer hubs; soft silicone tips
  • Sterile, packaged in pouches
  • Five needles per pouch, 20 pouches per box
Specifications
Specifications
0.078 in.
Yes
2 in.
PTFE
For critical applications requiring sterile, flexible, single use feeding needles
0.02cm
18g
0.5cm
Sterile
Soft, Flexible Animal feeding needle
Documents