Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
Manufacturer:  Cadence Science™ POP4184*
 View more versions of this product
Catalog No. 14-825-15X
Description
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
  • Stainless-steel needles with deflected noncoring septum
  • For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Specifications
20 G
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
Stainless steel
Deflected End
Deflected non-coring septum
2 in.
51mm
Penetration Needle
Documents