Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures.
Supplier: Cadence Science™ POP4187
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
- Penetration needles with deflected noncoring septum
- For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Nickel-plated Brass (Hub), Stainless Steel (Tube) | |
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures | |
8 in. | |
12 Needles |
20 G | |
Deflected non-coring septum | |
Penetration Needle | |
Deflected End |