Learn More
Cacodylic acid, sodium salt trihydrate, 98%, pure, Thermo Scientific Chemicals
Cacodylic acid, sodium salt trihydrate, CAS # 6131-99-3, is commonly used in research as a buffering agent in electron microscopy.
Supplier: Thermo Scientific Chemicals 214975000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Cacodylic acid sodium salt trihydrate is one of the major methylated metabolites of ingested arsenical compounds in most animals
- In in vivo experiments using rat model, cacodylic acid sodium salt trihydrate has been shown to promote cancer formation in the kidney, bladder, thyroid gland, and liver
Applications
- Cacodylic acid sodium salt trihydrate is used in research as a buffer during the preparation and fixation of biological samples for transmission in electron microscopy
- It has been used in buffers for formulating and optimizing screens
- In chemical experiments, this compound has been used to introduce arsenic into proteins for single-wavelength anomalous diffraction phasing in crystallography techniques
Specifications
Cacodylic acid, sodium salt trihydrate | |
6131-99-3 | |
98% | |
(CH3)2As(O)ONa·3H2O | |
15, 8730 | |
RLGWPHBPRCROJO-UHFFFAOYSA-M | |
sodium;dimethylarsinate;trihydrate | |
23679059 | |
≥97.5% | |
500 g | |
Glass bottle |
98% | |
White | |
C2H12AsNaO5 | |
MFCD00149079 | |
sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate | |
C[As](=O)(C)[O-].O.O.O.[Na+] | |
214.024 | |
214.03 | |
Pure | |
Authentic | |
Crystals or Crystalline Powder |
Chemical Identifiers
6131-99-3 | |
214.024 | |
RLGWPHBPRCROJO-UHFFFAOYSA-M | |
23679059 | |
C[As](=O)(C)[O-].O.O.O.[Na+] |
C2H12AsNaO5 | |
MFCD00149079 | |
sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate | |
sodium;dimethylarsinate;trihydrate |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
May cause cancer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RTECSNumber : CH7890000
RUO – Research Use Only