Learn More
Cacodylic acid sodium salt trihydrate 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1813914
Description
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.
Specifications
Cacodylic acid sodium salt trihydrate | |
C2H12AsNaO5 | |
MFCD00149079 | |
UN1688 | |
14,8595 | |
Soluble in water and alcohol. Insoluble in diethyl ether. | |
C[As](=O)(C)[O-].O.O.O.[Na+] | |
214.024 | |
214.03 (159.98 Anhydrous) |
6131-99-3 | |
(CH3)2AsO2Na·3H2O | |
25 g | |
3702348 | |
sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate | |
RLGWPHBPRCROJO-UHFFFAOYSA-M | |
sodium;dimethylarsinate;trihydrate | |
23679059 | |
≥98% |
Chemical Identifiers
6131-99-3 | |
214.024 | |
RLGWPHBPRCROJO-UHFFFAOYSA-M | |
23679059 | |
C[As](=O)(C)[O-].O.O.O.[Na+] |
C2H12AsNaO5 | |
MFCD00149079 | |
sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate | |
sodium;dimethylarsinate;trihydrate |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301+H331-H350-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SODIUM CACODYLATE
EINECSNumber : 204-708-2
RTECSNumber : CH7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only