Learn More
Cacodylic acid, 98%, Thermo Scientific Chemicals
$153.52 - $153.52
Chemical Identifiers
CAS | 75-60-5 |
---|---|
Molecular Formula | C2H7AsO2 |
Molecular Weight (g/mol) | 138.00 |
MDL Number | MFCD00002095 |
InChI Key | OGGXGZAMXPVRFZ-UHFFFAOYSA-N |
Synonym | cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 |
PubChem CID | 2513 |
ChEBI | CHEBI:48765 |
IUPAC Name | dimethylarsinic acid |
SMILES | C[As](C)(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC318150100
|
Thermo Scientific Chemicals
318150100 |
10 g | Glass bottle |
Each for $153.52
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-60-5 | |
138.00 | |
OGGXGZAMXPVRFZ-UHFFFAOYSA-N | |
2513 | |
dimethylarsinic acid |
C2H7AsO2 | |
MFCD00002095 | |
cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 | |
CHEBI:48765 | |
C[As](C)(O)=O |
Specifications
195°C to 198°C | |
White | |
C2H7AsO2 | |
MFCD00002095 | |
cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 | |
C[As](C)(O)=O | |
138.00 | |
CHEBI:48765 | |
≥97.5% | |
Authentic | |
Crystalline Powder |
75-60-5 | |
98% | |
(CH3)2As(O)OH | |
15, 1609 | |
OGGXGZAMXPVRFZ-UHFFFAOYSA-N | |
dimethylarsinic acid | |
2513 | |
138.01 | |
10 g | |
Glass bottle | |
Cacodylic acid |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only