Please login to your online account to display your discounted pricing

Alfa Aesar™ C3 Peptide P16

For cellular and molecular biology applications

$147.35 - $233.55

Specifications

Product Type C3 Peptide P16
Molecular Weight 1947.40
Storage Requirements -30° to -10°C
Format Solid
Sequence Lys-Asn-Arg-Trp-Glu-Asp-Pro-Gly-Lys-Gln-Leu-Tyr-Asn-Val-Glu-Ala
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66952EXD

 
alfa aesar™
J66952EXD
0.5mg Each for $147.35

AAJ66952MCR

 
alfa aesar™
J66952MCR
1mg Each for $233.55
Description & Specifications

Specifications

Product Type C3 Peptide P16
Molecular Weight 1947.40
Storage Requirements -30° to -10°C
Format Solid
Sequence Lys-Asn-Arg-Trp-Glu-Asp-Pro-Gly-Lys-Gln-Leu-Tyr-Asn-Val-Glu-Ala
Regulatory Status RUO
Purification Method HPLC