Please login to your online account to display your discounted pricing

Alfa Aesar™ C2 Dihydroceramide, 98%


Manufacturer: alfa aesar™  J67155LB0

 View more versions of this product

Catalog No. AAJ67155LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Assay 98%
Assay 98%
CAS 13031-64-6
Molecular Formula C20H41NO3
Formula Weight 343.55
Synonym D-erythro-N-Acetylsphinganine;
Quantity 5mg
Description White
Storage Requirements -30°C to -10°C
Format Solid
Type C2 Dihydroceramide, 98%