Please login to your online account to display your discounted pricing

Alfa Aesar™ C16 Ceramide, 98%


Manufacturer: alfa aesar™  J67093LB0

 View more versions of this product

Catalog No. AAJ67093LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

CAS 24696-26-2
Synonym D-erythro-N-Palmitoylsphingosine; N-Hexadecanoyl-D-erythro-sphingosine
Storage Requirements -30°C to -10°C
Type C16 Ceramide, 98%
Molecular Formula C34H67NO3
For Use With (Application) An endogenous ceramide that acts as a lipid second messenger to regulate apoptosis and stress signaling.
Formula Weight 537.91
Solubility Soluble in DMF at 0.15mg/ml
Format Crystalline solid
Quantity 5mg
Assay 98%