Please login to your online account to display your discounted pricing

Alfa Aesar™ C-Type Natriuretic Peptide, Chicken

For cellular and molecular biology applications

Specifications

Product Type C-Type Natriuretic Peptide, chicken
Cross Reactivity Chicken
Molecular Weight 2241.61
Storage Requirements -30° to -10°C
Sequence Gly-Leu-Ser-Arg-Ser-Cys-Phe-Gly-Val-Lys-Leu-Asp-Arg- Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bridge:Cys6-Cys22)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66410EXC

 
alfa aesar™
J66410EXC
2.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66410MCR

 
alfa aesar™
J66410MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type C-Type Natriuretic Peptide, chicken
Cross Reactivity Chicken
Molecular Weight 2241.61
Storage Requirements -30° to -10°C
Sequence Gly-Leu-Ser-Arg-Ser-Cys-Phe-Gly-Val-Lys-Leu-Asp-Arg- Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bridge:Cys6-Cys22)
Format Solid
Regulatory Status RUO
Purification Method HPLC