Please login to your online account to display your discounted pricing

c-Rel Rat anti-Mouse, PE, Clone: 1RELAH5, eBioscience™

Rat Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen c-Rel
Clone 1RELAH5
Host Species Rat
Gene Alias Rel
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245513

 
life technologies
LS12611180
25μg Each for $216.05

50245514

 
life technologies
LS12611182
100μg Each for $506.05
Description & Specifications

Specifications

Antigen c-Rel
Clone 1RELAH5
Host Species Rat
Gene Alias Rel
Species Reactivity Mouse
Applications Flow Cytometry
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 1RELAH5 monoclonal antibody reacts with mouse c-Rel. c-Rel is a 65kDa hematopoietic lineage-restricted member of the NF-kB family of transcription factors. In the thymus, c-Rel is upregulated in regulatory T cells (Treg) when compared to other CD4+ thymocytes. c-Rel is believed to play a critical role in thymic Treg (nTreg) development, but is not necessary for TGF beta-mediated T reg (iTreg) differentiation. C-Rel is also involved in proliferation and survival in mature T cells.