missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

c-Rel Rat anti-Mouse, eFluor 660, Clone: 1RELAH5, eBioscience™

Rat Monoclonal Antibody

$196.68 - $487.08

Specifications

Antigen c-Rel
Clone 1RELAH5
Host Species Rat
Gene Alias Rel
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124416 Life Technologies
LS50611180
25μg Each for $196.68
501124417 Life Technologies
LS50611182
100μg Each for $487.08
Specifications

Description

The 1RELAH5 monoclonal antibody reacts with mouse c-Rel. c-Rel is a 65kDa hematopoietic lineage-restricted member of the NF-kB family of transcription factors. In the thymus, c-Rel is upregulated in regulatory T cells (Treg) when compared to other CD4+ thymocytes. c-Rel is believed to play a critical role in thymic Treg (nTreg) development, but is not necessary for TGF beta-mediated T reg (iTreg) differentiation. C-Rel is also involved in proliferation and survival in mature T cells.
Description & Specifications

Specifications

c-Rel
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1RELAH5
Rel
Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL