Please login to your online account to display your discounted pricing

Alfa Aesar™ C-Reactive Protein

For research use

$221.50 - $221.50

Specifications

Formulation Buffered solution
Recombinant No
Product Type C-Reactive Protein
Format Liquid
For Use With (Application) For immunological applications in the preparation of antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63028MCR

 
alfa aesar™
J63028MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ63028AMH

 
alfa aesar™
J63028AMH
200μg Each for $221.50
Description & Specifications

Specifications

Formulation Buffered solution
Recombinant No
Product Type C-Reactive Protein
Format Liquid
For Use With (Application) For immunological applications in the preparation of antisera
Regulatory Status RUO