missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ C-Peptide, Dog

For cellular and molecular biology applications

Specifications

Product Type C-Peptide, dog
Cross Reactivity Dog
Molecular Weight 3174.46
Storage Requirements -30° to -10°C
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66573MCR
View Documents
Alfa Aesar™
J66573MCR
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66573EXD
View Documents
Alfa Aesar™
J66573EXD
0.5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

C-Peptide, dog
3174.46
Solid
HPLC
Dog
-30° to -10°C
RUO

For Research Use Only