missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ C-Myc Peptide
For cellular and molecular biology applications
Supplier: Thermo Scientific™ J66846MCR
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
HPLC | |
Solid | |
C-Myc Peptide | |
RUO - research use only |
-30°C to -10°C | |
1203.29 | |
1 mg | |
Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu |
RUO â Research Use Only