Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ c-myc Ab-5, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

$311.22 - $1,289.29

Specifications

Antigen c-myc Ab-5
Clone 67P05
Concentration 200μg/mL
Applications Immunofluorescence
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1054P

 
thermo scientific™ lab vision™
MS1054P
200μg/mL 1mL Each for $1,289.29

MS1054P0

 
thermo scientific™ lab vision™
MS1054P0
200μg/mL 100μL Each for $311.22

MS1054P1

 
thermo scientific™ lab vision™
MS1054P1
200μg/mL 500μL Each for $802.08
Description & Specifications

Specifications

Antigen c-myc Ab-5
Clone 67P05
Concentration 200μg/mL
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Transcription Regulation
Type Primary
Immunogen Recombinant c-myc protein
Isotype IgG1

Obtain accurate and reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ c-myc Ab-5, Mouse Monoclonal Antibody.

c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Translocation of the c-myc locus on chromosome 8 to the immunoglobulin loci on chromosome 14 (heavy chain); 2 (δ light chain); or 22 (λ light chain) is described in Burkitt's lymphoma and other B-cell lymphoproliferative conditions. An aberrant expression of the c-myc gene occurs in tumors of different origins such as colorectal, gastric, gallbladder, hepatic, mammary, ovarian, endometrial, head and neck, pulmonary, prostatic, thyroidal, oral, ocular, nasopharyngeal, endocrine, as well as hematopoietic neoplasms.

Host Species: Mouse

Clone: 67P05

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat and Chicken. Others not tested

Epitope: Not determined

Immunogen: Recombinant c-myc protein

Molecular Weight: 64-67 kDa

Positive Control: Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and Denatured)
  • Gel Supershift (Not verified)

Note:

USA: RUO; Int'l: RUO