missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ c-myc Ab-1, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunoprecipitation experiments with Thermo Scientific™ c-myc Ab-1, Mouse Monoclonal Antibody.

$246.33 - $1,217.33

Specifications

Antigen c-myc Ab-1
Clone 9E11; same as A3B2
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS127P Thermo Scientific™ Lab Vision™
MS127P
200μg/mL 1mL Each for $1,154.02
MS127P0 Thermo Scientific™ Lab Vision™
MS127P0
200μg/mL 100μL Each for $246.33
MS127P1 Thermo Scientific™ Lab Vision™
MS127P1
200μg/mL 500μL Each for $717.14
MS127P1ABX Thermo Scientific™ Lab Vision™
MS127P1ABX
1mg/mL 100μL Each for $812.71
MS127PABX Thermo Scientific™ Lab Vision™
MS127PABX
1mg/mL 200μL Each for $1,217.33
Specifications

Description

Ensure accurate and reproducible results in immunoprecipitation experiments with Thermo Scientific™ c-myc Ab-1, Mouse Monoclonal Antibody.

c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Translocation of the c-myc locus on chromosome 8 to the immunoglobulin loci on chromosome 14 (heavy chain); 2 (δ light chain); or 22 (light chain) is described in Burkitt's lymphoma and other B-cell lymphoproliferative conditions. An aberrant expression of the c-myc gene occurs in tumors of different origins such as colorectal, gastric, gallbladder, hepatic, mammary, ovarian, endometrial, head and neck, pulmonary, prostatic, thyroidal, oral, ocular, nasopharyngeal, endocrine, as well as hematopoietic neoplasms.

Host Species: Mouse

Clone: 9E11; same as A3B2

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse and Chicken. Others not known.

Epitope: aa 408-439 (Different from Ab-2)

Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc

Molecular Weight: 64-67 kDa

Positive Control: Raji cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunoprecipitation (Native and denatured)
  • Epitope Tagging (Not Suitable)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

c-myc Ab-1
Immunoprecipitation
Unlabeled
Concentrated
RUO
Primary
IgG2a kappa
9E11; same as A3B2
Monoclonal
Chicken,Human,Mouse,Chicken,Human,Mouse,Chicken,Human,Mouse
Murine
Transcription Regulation
A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc