missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.

$269.82 - $1,372.92

Specifications

Antigen c-myb/v-myb Ab-1
Clone 4E3
Applications Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1728P Thermo Scientific™ Lab Vision™
MS1728P
Purified Ab with BSA and Azide 1mL Each for $1,263.43
MS1728P0 Thermo Scientific™ Lab Vision™
MS1728P0
Purified Ab with BSA and Azide 100μL Each for $269.82
MS1728P1 Thermo Scientific™ Lab Vision™
MS1728P1
Purified Ab with BSA and Azide 500μL Each for $756.41
MS1728P1ABX Thermo Scientific™ Lab Vision™
MS1728P1ABX
Ab wthout BSA/Azide (Frozen) 100μL Each for $826.02
MS1728PABX Thermo Scientific™ Lab Vision™
MS1728PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,372.92
MS1728R7 Thermo Scientific™ Lab Vision™
MS1728R7
Ready-to-use for IHC (UVLP) 7mL Each for $642.10
Specifications

Description

Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.

Transcription factor Myb is expressed in hematopoietic cells and immature gastroepithelial cells.

Host Species: Mouse

Clone: 4E3

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant v-myb protein

Molecular Weight: ∽80 kDa

Positive Control: Jurkat and Ls174T cells, Testis

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

c-myb/v-myb Ab-1
Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Unlabeled
RUO
Primary
IgG2a
4E3
Monoclonal
Murine
Transcription Regulation
Recombinant v-myb protein