Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody


Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1728PABX

 View more versions of this product

Catalog No. MS1728PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Isotype IgG2a
Antigen c-myb/v-myb Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Recombinant v-myb protein
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Clone 4E3
Quantity 200µL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Transcription Regulation
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate and reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ c-myb/v-myb Ab-1, Mouse Monoclonal Antibody.

Transcription factor Myb is expressed in hematopoietic cells and immature gastroepithelial cells.

Host Species: Mouse

Clone: 4E3

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant v-myb protein

Molecular Weight: ∽80 kDa

Positive Control: Jurkat and Ls174T cells, Testis

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO