missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

c-Met (HGF Receptor) Rat anti-Mouse, PE, Clone: eBioclone 7, eBioscience™

Rat Monoclonal Antibody

$157.08 - $407.88

Specifications

Antigen c-Met (HGF Receptor)
Clone eBioclone 7
Host Species Rat
Gene Alias hepatocyte growth factor receptor
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011223 Life Technologies
LS12885480
25μg Each for $157.08
5011224 Life Technologies
LS12885482
100μg Each for $407.88
Specifications

Description

The eBioclone 7 monoclonal antibody was generated against a mouse c-Met-Ig fusion protein, and reacts with mouse c-Met-transfected cells. Mouse c-Met (HGFR) is a 170kDa receptor tyrosine kinase (RTK) expressed by epithelial cells of the brain, kidney, liver and other tissues. Binding of its ligand, Hepatocyte Growth Factor (HGF), triggers receptor autophosphorylation, and activation of several downstream effectors including the mitogen-activated protein kinases ERK-1 and ERK-2, and PLC gamma. Activation of the c-Met signal transduction pathway leads to mulitple cellular responses including cell motility, scattering, proliferation, survival and angiogenesis. Mutations in human c-Met have been implicated in the development of several malignancies.
Description & Specifications

Specifications

c-Met (HGF Receptor)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioclone 7
hepatocyte growth factor receptor
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL