Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ c-erbB-3/HER-3 Ab-6, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ c-erbB-3/HER-3 Ab-6, Mouse Monoclonal Antibody.

$277.12 - $1,554.00

Specifications

Antigen HER-3/c-erbB-3 Ab-6
Clone 2B5
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS310B

 
thermo scientific™ lab vision™
MS310B
200μg/mL Biotin 1mL Each for $1,554.00

MS310B0

 
thermo scientific™ lab vision™
MS310B0
200μg/mL Biotin 100µL Each for $370.88

MS310B1

 
thermo scientific™ lab vision™
MS310B1
200μg/mL Biotin 500µL Each for $1,021.10

MS310P

 
thermo scientific™ lab vision™
MS310P
200μg/mL Unlabeled 1mL Each for $1,236.06

MS310P0

 
thermo scientific™ lab vision™
MS310P0
200μg/mL Unlabeled 100µL Each for $277.12

MS310P1

 
thermo scientific™ lab vision™
MS310P1
200μg/mL Unlabeled 500µL Each for $779.71

MS310P1ABX

 
thermo scientific™ lab vision™
MS310P1ABX
1mg/mL Unlabeled 100µL Each for $848.39

MS310PABX

 
thermo scientific™ lab vision™
MS310PABX
1mg/mL Unlabeled 200µL Each for $1,289.82
Description & Specifications

Specifications

Antigen HER-3/c-erbB-3 Ab-6
Clone 2B5
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Recombinant rat c-erbB-3/HER-3 oncoprotein
Isotype IgG1

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ c-erbB-3/HER-3 Ab-6, Mouse Monoclonal Antibody.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2B5

Isotype: IgG1

Species Reactivity: Human, Rat, Mouse. Others not tested.

Epitope: aa 1234-1259

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D or MAD109 cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO