Please login to your online account to display your discounted pricing

C-1, Tocris


Protein kinase C inhibitor

Quantity 10mg

Manufacturer: r & d systems  054310

 View more versions of this product

Catalog No. 054310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 277.34
CAS 84468-24-6
Assay Percent Range >99%
Chemical Name or Material C-1
Molecular Formula C13H15N3O2S
Synonym 1-(5-Isoquinolinesulfonyl)piperazine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material C-1
Assay Percent Range >99%
Molecular Formula C13H15N3O2S
Formula Weight 277.34
Synonym 1-(5-Isoquinolinesulfonyl)piperazine
Accessories

Store at Room Temperature