Please login to your online account to display your discounted pricing

C-1, Tocris

Protein kinase C inhibitor

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material C-1
Assay Percent Range >99%
Molecular Formula C13H15N3O2S
Formula Weight 277.34
Synonym 1-(5-Isoquinolinesulfonyl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

054310

 
r & d systems
054310
10mg Each for $192.67

054350

 
r & d systems
054350
50mg Each for $812.12
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 277.34
Chemical Name or Material C-1
Assay Percent Range >99%
Molecular Formula C13H15N3O2S
CAS 84468-24-6
Synonym 1-(5-Isoquinolinesulfonyl)piperazine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature