Please login to your online account to display your discounted pricing

Tocris™ C 021 dihydrochloride

Potent CCR4 antagonist

$252.39 - $1,050.42

Specifications

Product Type C 021 dihydrochloride
Inhibitors C 021 dihydrochloride
Molecular Weight 540.57
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

358110R

 
tocris™
358110
10mg Each for $252.39

358150R

 
tocris™
358150
50mg Each for $1,050.42
Description & Specifications

Specifications

Product Type C 021 dihydrochloride
Inhibitors C 021 dihydrochloride
Molecular Weight 540.57
Purity >99

Potent CCR4 chemokine receptor antagonist (IC50 values are 0.14 and 0.039μM for inhibition of chemotaxis in human and mouse respectively).

  • Chemical Name: 2-[1,4'-Bipiperidin]-1'-yl-N-cycloheptyl-6,7-dimethoxy-4-quinazolinamine dihydrochloride
  • Formula: C27H41N5O2.2HCl
  • CAS Number: 864289-85-0
  • Solubility: Soluble to 50mM in water and to 100mM in DMSO
  • Purity: >99