Please login to your online account to display your discounted pricing

Tocris™ BYK 204165

Selective PARP-1 inhibitor

$205.70 - $862.95

Specifications

Product Type BYK 204165
Inhibitors BYK 204165
Molecular Weight 252.27
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

373410R

 
tocris™
RDS373410
10mg Each for $205.70

373450R

 
tocris™
RDS373450
50mg Each for $862.95
Description & Specifications

Specifications

Product Type BYK 204165
Inhibitors BYK 204165
Molecular Weight 252.27
Purity >99

Potent and selective poly(ADP-ribose) polymerase (PARP)-1 inhibitor (pIC50 values are 5.38 and 7.35 for PARP-2 and PARP-1 respectively).

  • Chemical Name: 4-[(1-Methyl-1H-pyrrol-2-yl)methylene]-1,3(2H,4H)-isoquinolinedione
  • Formula: C15H12N2O2
  • CAS Number: 1104546-89-5
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99