Please login to your online account to display your discounted pricing

BW-B 70C, Tocris


5-Lipoxygenase inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS130410

 View more versions of this product

Catalog No. 130410

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 316.33
Quantity 10mg
Molecular Formula C17H17FN2O3
CAS 134470-38-5
Synonym N-[3-[3-(-Fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxyurea
Recommended Storage Desiccate at -20°C
Chemical Name or Material BW-B 70C
Assay Percent Range >98%

Specifications

Chemical Name or Material BW-B 70C
Assay Percent Range >98%
Molecular Formula C17H17FN2O3
Formula Weight 316.33
Synonym N-[3-[3-(-Fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxyurea
Accessories

Desiccate at -20°C