Please login to your online account to display your discounted pricing

BW-B 70C, Tocris

5-Lipoxygenase inhibitor

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material BW-B 70C
Assay Percent Range >98%
Molecular Formula C17H17FN2O3
Formula Weight 316.33
Synonym N-[3-[3-(-Fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxyurea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

130410

 
r & d systems
RDS130410
10mg Each for $214.12

130450

 
r & d systems
RDS130450
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 316.33
Quantity 10mg
CAS 134470-38-5
Recommended Storage Desiccate at -20°C
Molecular Formula C17H17FN2O3
Synonym N-[3-[3-(-Fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxyurea
Chemical Name or Material BW-B 70C
Assay Percent Range >98%
Safety and Handling

Desiccate at -20°C