Please login to your online account to display your discounted pricing

Alfa Aesar™ Butyrylferrocene, 99%

$33.76 - $33.76

Chemical Identifiers

Chemical Name or Material Butyrylferrocene
CAS 1271-94-9
Formula Weight 256.13
MDL Number MFCD00143883
Synonym Butyroferrocene; Ferrocenyl n-propyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3940404

  SDS
alfa aesar™
3940404
2g Each for $33.76

AA3940409

  SDS
alfa aesar™
3940409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.13
Chemical Name or Material Butyrylferrocene
Physical Form Solid
Assay 99%
MDL Number MFCD00143883
CAS 1271-94-9
Synonym Butyroferrocene; Ferrocenyl n-propyl ketone
Density 1.254g/mL
Boiling Point 329°C
Melting Point 36° to 40°C
Quantity 2g