missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Butyrylferrocene, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 3940409
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Butyrylferrocene | |
36°C to 40°C | |
329°C | |
MFCD00143883 | |
Butyroferrocene; Ferrocenyl n-propyl ketone | |
[Fe].c1cccc1.CCCC(=O)c1cccc1 | |
256.13 | |
99% |
1271-94-9 | |
1.254 g/mL | |
C14H16FeO | |
10 g | |
CVNYMHASFXAXJV-UHFFFAOYNA-N | |
Butyrylferrocene | |
256.13 | |
Solid |
Chemical Identifiers
1271-94-9 | |
256.13 | |
CVNYMHASFXAXJV-UHFFFAOYNA-N | |
Butyrylferrocene |
C14H16FeO | |
MFCD00143883 | |
Butyroferrocene; Ferrocenyl n-propyl ketone | |
[Fe].c1cccc1.CCCC(=O)c1cccc1 |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 215-045-3
TSCA : No
Recommended Storage : Ambient temperatures